Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026763.1 | 1 | 1295 | |||
GenBank mRNA | AF071111.1 | - | 1 | 1295 | ||
AF086556.1 | - | 1 | 51 |