Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002925.2 | 1 | 34 | |||
GenBank mRNA | BC014341.1 | - | 1 | 38 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX283830.1 | - | 1 | 35 | Chr. 1 - 166603916 - 166620871 [+] on Build GRCh38 |