Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOB2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002936.2 | 1 | 50 | |||
GenBank mRNA | BC015543.1 | - | 1 | 70 | ||
BC050562.1 | - | 1 | 50 |