Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028077.1 | 6 | 3215 | |||
NR_133668.1 | 3 | 1453 | ||||
NR_133669.1 | 3 | 1518 | ||||
NR_136509.1 | 6 | 3393 | ||||
NR_136510.1 | 7 | 3509 | ||||
NR_136511.1 | 5 | 2068 | ||||
NR_136512.1 | 6 | 3427 | ||||
NR_136513.1 | 3 | 1453 | ||||
XM_011515014.2 | XP_011513316.1 | 7 | 6359 | |||
XM_011515015.2 | XP_011513317.1 | 6 | 10483 | |||
XM_011515017.2 | XP_011513319.1 | 7 | 6181 | |||
XM_017011528.1 | XP_016867017.1 | 7 | 3468 | |||
GenBank mRNA | AB007886.1 | - | 5 | 4477 | ||
BC041661.1 | - | 6 | 3179 | 6063526 |