Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097775.1 | - | 3 | 632 |
Gene Symbol: CHKB-CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB051457.1 | - | 1 | 1016 |