Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003501.1 | 9, 10 | 2464 | |||
GenBank mRNA | AB075850.1 | - | 9, 10 | 2421 | ||
AL832489.1 | - | 9, 10 | 2434 | |||
BX648129.1 | - | 7, 8 | 2137 |