Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM225B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024376.1 | 3 | 5286 | |||
GenBank mRNA | AL045730.1 | - | 1 | 35 |
Gene Symbol: FAM225A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125961.1 | - | 1 | 2610 | ||
CR627046.1 | - | 2 | 5476 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA399228.1 | - | 1 | 432 | Chr. 9 - 113105092 - 113105673 [-] on Build GRCh38 |