Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM225B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024376.1 | 3 | 5428 | |||
GenBank mRNA | AL045730.1 | - | 1 | 177 |
Gene Symbol: FAM225A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125961.1 | - | 1 | 2752 | ||
CR627046.1 | - | 2 | 5618 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | T19103.1 | - | 1 | 262 | Chr. 2 - 231713176 - 231713541 [+] on Build GRCh38 |