Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256006.1 | NP_001242935.1 | 3 | 191 | ||
NR_027459.2 | 3 | 191 | ||||
XM_017000932.1 | XP_016856421.1 | 3 | 181 | |||
GenBank mRNA | AJ617624.1 | - | 2 | 72 | ||
AJ617625.1 | - | 3 | 105 |