Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024281.1 | 1 | 2550 | |||
GenBank mRNA | AF052108.1 | - | 1 | 970 | ||
AK055056.1 | - | 1 | 1466 | |||
AK091593.1 | - | 1 | 2299 | |||
BC042097.1 | - | 1 | 421 |
Gene Symbol: LRRC37A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308554.1 | - | 1675 |