Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024281.1 | 2 | 2682 | |||
GenBank mRNA | AF052108.1 | - | 1 | 1178 | ||
AK055056.1 | - | 1 | 1674 | |||
AK091593.1 | - | 1 | 2507 | |||
AL832535.1 | - | 2 | 1407 | |||
BC042097.1 | - | 1 | 629 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM689652.1 | - | 1 | 179 | Chr. 8 - 9900154 - 9900742 [-] on Build GRCh38 |