Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL18A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027498.1 | 3 | 2148 | |||
NR_028082.1 | 3 | 3232 | ||||
GenBank mRNA | AK096071.1 | - | 3 | 2148 | ||
BC111539.2 | - | 3 | 3232 | 40026216 | ||
BC111540.2 | - | 3 | 2118 | 40026217 |