Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RACGAP1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026583.1 | 1 | 1969 | |||
GenBank mRNA | AF334184.1 | - | 1 | 1977 | ||
BC125190.1 | - | 1 | 1960 | |||
BC125191.1 | - | 1 | 1960 |