Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7E47P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120437.1 | 3 | 1213 | |||
NR_120439.1 | 2 | 736 | ||||
NR_120440.1 | 2 | 713 | ||||
GenBank mRNA | BC042060.1 | - | 3 | 1213 | ||
DB348459.1 | - | 1 | 243 |
Gene Symbol: OR7E12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_044999.1 | 1 | 874 |
Gene Symbol: OR7E37P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002163.3 | 1 | 864 | |||
GenBank mRNA | AA235800.1 | - | 1 | 251 |