Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293626.1 | NP_001280555.1 | 47 | 5287 | ||
XM_011516692.2 | XP_011514994.1 | 40 | 4502 | |||
XM_011516693.2 | XP_011514995.1 | 26 | 2767 | |||
XM_011516694.2 | XP_011514996.1 | 17 | 1628 |