Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002009.2 | NP_001002009.1 | 1, 2 | 232 | ||
GenBank mRNA | BC066914.1 | - | 1, 2 | 157 | 5264738 | |
CB137440.1 | - | 1, 2 | 229 |