Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278298.1 | NP_001265227.1 | 40 | 8289 | ||
NR_022012.2 | 40 | 8289 | ||||
XM_011512621.2 | XP_011510923.1 | 39 | 8100 | |||
GenBank mRNA | AK093199.1 | - | 6 | 1509 | ||
AM906083.1 | - | 6 | 1623 | |||
AM906084.1 | - | 3 | 861 | |||
EU085556.1 | - | 40 | 8289 |