Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022012.2 | 41 | 8341 | |||
XM_011512621.2 | XP_011510923.1 | 40 | 8152 | |||
GenBank mRNA | AK093199.1 | - | 7 | 1561 | ||
AM906084.1 | - | 4 | 913 | |||
EU085556.1 | - | 41 | 8341 |