Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026707.1 | 4 | 1257 | |||
NR_026708.1 | 4 | 1257 | ||||
NR_026709.1 | 3 | 1194 | ||||
GenBank mRNA | AK023750.1 | - | 2 | 978 | ||
AY042235.1 | - | 4 | 1139 | |||
AY042236.1 | - | 4 | 1139 | |||
AY042239.1 | - | 3 | 1076 |