Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015413.1 | 6 | 781 | |||
NR_024410.1 | 5 | 711 | ||||
NR_024411.1 | 4 | 600 | ||||
GenBank mRNA | BC033338.1 | - | 6 | 713 | ||
BC106956.2 | - | 5 | 515 | |||
BC106957.2 | - | 4 | 404 | |||
BC113009.1 | - | 4 | 404 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB508949.2 | - | 3 | 355 | Chr. 10 - 10934950 - 10946576 [-] on Build GRCh38 |