Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028052.1 | 1 | 523 | |||
GenBank mRNA | AB029315.1 | - | 1 | 521 | ||
AF225423.1 | - | 1 | 412 | |||
AF242770.1 | - | 1 | 477 | |||
AK312641.1 | - | 2 | 326 | |||
AL355700.1 | - | 1 | 473 | |||
AL360142.1 | - | 1 | 495 | |||
AL834503.1 | - | 1 | 512 | |||
BC007222.1 | - | 1 | 498 | 2819984 | ||
BC009389.2 | - | 1 | 473 | 4130266 | ||
BC017192.2 | - | 1 | 478 | 3139180 | ||
BC019023.2 | - | 1 | 479 | |||
BG716404.1 | - | 1 | 528 |