Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM95B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026759.1 | 5 | 1511 | |||
GenBank mRNA | AK093443.1 | - | 1 | 1873 | ||
AK094938.1 | - | 1 | 2810 | |||
AK125850.1 | - | 4 | 4208 | |||
AL833349.1 | - | 1 | 4659 |
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093059.1 | - | 8 | 2396 |
Gene Symbol: FAM95A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038409.1 | 3 | 2365 | |||
GenBank mRNA | AL137712.1 | - | 3 | 2365 |
Gene Symbol: FAM95C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047651.1 | 3 | 2640 | |||
GenBank mRNA | AK097951.1 | - | 3 | 2642 | ||
AL834388.1 | - | 1 | 893 |
Gene Symbol: LOC105379252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136310.1 | 5 | 1511 |