Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027397.1 | 2, 3 | 519 | |||
XM_006711628.3 | XP_006711691.1 | 4, 5 | 790 | |||
XM_011510150.2 | XP_011508452.1 | 3, 4 | 693 | |||
XM_011510151.1 | XP_011508453.1 | 2, 3 | 442 | |||
XM_017002807.1 | XP_016858296.1 | 4, 5 | 756 | |||
GenBank mRNA | AK308339.1 | - | 2, 3 | 519 |