Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257205.1 | NP_001244134.1 | 1 | 233 | ||
NM_001257206.1 | NP_001244135.1 | 1 | 233 | |||
GenBank mRNA | AK289762.1 | - | 1 | 173 | ||
DC344309.1 | - | 1 | 233 | |||
DQ655917.2 | - | 1 | 215 | |||
M60515.1 | - | 1 | 107 |