Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC75A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027158.1 | 1 | 44 | |||
NR_027159.1 | 1 | 44 | ||||
NR_027160.1 | 1 | 44 | ||||
NR_027161.1 | 1 | 44 | ||||
NR_027162.1 | 1 | 44 | ||||
NR_027163.1 | 1 | 44 | ||||
NR_027164.1 | 1 | 44 | ||||
NR_027165.1 | 1 | 44 | ||||
NR_027166.1 | 1 | 44 | ||||
NR_027167.1 | 1 | 44 | ||||
NR_027169.1 | 1 | 44 | ||||
NR_027170.1 | 1 | 44 | ||||
NR_027171.1 | 1 | 44 | ||||
NR_027173.1 | 1 | 44 | ||||
NR_027174.1 | 1 | 44 | ||||
NR_027175.1 | 1 | 44 | ||||
NR_027176.1 | 1 | 44 | ||||
NR_027177.1 | 1 | 44 | ||||
NR_027178.1 | 1 | 44 | ||||
NR_027179.1 | 1 | 44 | ||||
NR_027667.1 | 1 | 44 | ||||
NR_045021.1 | 1 | 44 | ||||
NR_045022.1 | 1 | 44 | ||||
NR_045023.1 | 1 | 44 | ||||
NR_045024.1 | 1 | 44 | ||||
NR_045025.1 | 1 | 44 | ||||
NR_045026.1 | 1 | 44 | ||||
NR_045027.1 | 1 | 44 | ||||
NR_045028.1 | 1 | 44 | ||||
NR_045029.1 | 1 | 44 | ||||
GenBank mRNA | CN297624.1 | - | 1 | 13 |