Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026969.1 | 4 | 823 | |||
GenBank mRNA | BC039371.1 | - | 6 | 2125 | ||
BC101702.1 | - | 2 | 198 | |||
BC101704.1 | - | 2 | 197 |