Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002798.1 | 9 | 1303 | |||
GenBank mRNA | AF090387.1 | - | 9 | 1310 | ||
AF098485.1 | - | 9 | 1310 | |||
AF200344.1 | - | 9 | 1311 | |||
BC085008.1 | - | 9 | 1256 | |||
BC111705.1 | - | 9 | 1300 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF662964.1 | - | 2 | 531 | Chr. 19 - 50333796 - 50334561 [-] on Build GRCh38 |