Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002798.1 | 7 | 864 | |||
GenBank mRNA | AF090387.1 | - | 7 | 871 | ||
AF098485.1 | - | 7 | 871 | |||
AF200344.1 | - | 7 | 872 | |||
AK300884.1 | - | 7 | 875 | |||
AK310134.1 | - | 7 | 882 | |||
BC085008.1 | - | 7 | 817 | |||
BC111705.1 | - | 7 | 861 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ059485.1 | - | 5 | 578 | Chr. 19 - 50333923 - 50362024 [-] on Build GRCh38 |