Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002798.1 | 8 | 1013 | |||
GenBank mRNA | AF090387.1 | - | 8 | 1020 | ||
AF098485.1 | - | 8 | 1020 | |||
AF200344.1 | - | 8 | 1021 | |||
BC085008.1 | - | 8 | 966 | |||
BC111705.1 | - | 8 | 1010 |