Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAS-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002785.2 | 2 | 394 | |||
GenBank mRNA | AJ251759.1 | - | 2 | 394 | ||
DB067618.1 | - | 2 | 370 |