Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAS-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002785.2 | 3, 4 | 520 | |||
GenBank mRNA | AJ251759.1 | - | 3, 4 | 520 |