Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024182.1 | 4 | 408 | |||
NR_024183.1 | 3 | 536 | ||||
NR_024184.1 | 3 | 536 | ||||
GenBank mRNA | AK097741.1 | - | 3 | 531 | ||
BC021728.2 | - | 3 | 535 | |||
BC031252.1 | - | 4 | 408 | |||
BI458986.1 | - | 3 | 540 | |||
DB446670.1 | - | 4 | 288 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI463117.1 | - | 4 | 413 | Chr. 14 - 93997268 - 94001366 [+] on Build GRCh38 |