Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024182.1 | 2 | 49 | |||
NR_024183.1 | 2 | 301 | ||||
NR_024184.1 | 2 | 301 | ||||
GenBank mRNA | AK097741.1 | - | 2 | 296 | ||
BC021728.2 | - | 2 | 300 | |||
BC031252.1 | - | 2 | 49 | |||
BI458986.1 | - | 2 | 305 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI463117.1 | - | 2 | 54 | Chr. 14 - 93997268 - 94001366 [+] on Build GRCh38 |