Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006635.2 | NP_001006636.1 | 2 | 444 | ||
GenBank mRNA | AK303564.1 | - | 2 | 369 | ||
AK315918.1 | - | 2 | 380 | |||
BC017271.2 | - | 2 | 374 | 4635884 | ||
BG699784.1 | - | 2 | 445 |