Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024090.2 | 8 | 1582 | |||
NR_109786.1 | 8 | 1693 | ||||
NR_109787.1 | 7 | 1458 | ||||
NR_109788.1 | 7 | 1519 | ||||
GenBank mRNA | AK094858.1 | - | 7 | 1407 | ||
BC042856.1 | - | 8 | 1537 | |||
BC044727.1 | - | 8 | 1537 | |||
BC047405.1 | - | 8 | 1655 | |||
BX537735.1 | - | 7 | 1519 |