Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXT-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028119.1 | 2 | 392 | |||
GenBank mRNA | AK295794.1 | - | 1 | 1086 | ||
DB182705.1 | - | 2 | 393 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB136856.1 | - | 2 | 373 | Chr. X - 47658853 - 47660056 [+] on Build GRCh38 |