Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003086.1 | 3 | 335 | |||
GenBank mRNA | AF165514.1 | - | 4 | 381 | ||
AF165517.1 | - | 3 | 331 | |||
AK293968.1 | - | 3 | 328 | |||
AY829267.1 | - | 3 | 344 | |||
BC071987.1 | - | 3 | 335 |