Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003087.1 | 3 | 468 | |||
NR_003088.1 | 3 | 468 | ||||
GenBank mRNA | AF418600.1 | - | 3 | 468 | ||
AF518320.1 | - | 3 | 237 | |||
AF518321.1 | - | 3 | 323 | |||
AY063514.1 | - | 3 | 348 | |||
AY063515.1 | - | 3 | 387 | |||
AY344117.1 | - | 3 | 376 | |||
BC144077.1 | - | 2 | 201 | |||
DR004370.1 | - | 3 | 479 |