Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRA1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028045.1 | 1, 2 | 306 | |||
GenBank mRNA | AF047445.1 | - | 1, 2 | 36 | ||
AK296103.1 | - | 1, 2 | 43 | |||
AK304677.1 | - | 1, 2 | 92 | |||
AK310151.1 | - | 1, 2 | 140 | |||
BC101471.1 | - | 1, 2 | 23 | 8068977 | ||
BC101473.1 | - | 1, 2 | 26 | 8068979 | ||
DA776936.1 | - | 1, 2 | 306 |