Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRA1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028045.1 | 5 | 949 | |||
GenBank mRNA | AF047445.1 | - | 5 | 679 | ||
AK290077.1 | - | 4 | 650 | |||
AK296103.1 | - | 5 | 624 | |||
AK304677.1 | - | 4 | 645 | |||
AK310151.1 | - | 3 | 456 | |||
AY334570.1 | - | 3 | 493 | |||
BC069672.1 | - | 5 | 652 | 7262588 | ||
BC069697.1 | - | 5 | 652 | 7262600 | ||
BC069734.1 | - | 5 | 652 | 7262612 | ||
BC101471.1 | - | 5 | 666 | 8068977 | ||
BC101473.1 | - | 5 | 669 | 8068979 |