Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRA1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028045.1 | 6 | 1079 | |||
GenBank mRNA | AK290077.1 | - | 5 | 780 | ||
AK296103.1 | - | 6 | 754 | |||
AK304677.1 | - | 5 | 775 | |||
AK310151.1 | - | 4 | 586 | |||
BC069672.1 | - | 6 | 782 | 7262588 | ||
BC069697.1 | - | 6 | 781 | 7262600 | ||
BC069734.1 | - | 6 | 782 | 7262612 |