Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003099.1 | 2 | 242 | |||
XM_017011689.1 | XP_016867178.1 | 6 | 6167 | |||
XM_017011690.1 | XP_016867179.1 | 5 | 6078 | |||
XM_017011691.1 | XP_016867180.1 | 6 | 3369 | |||
XM_017011692.1 | XP_016867181.1 | 4 | 5553 | |||
XM_017011693.1 | XP_016867182.1 | 5 | 1259 | |||
XM_017011694.1 | XP_016867183.1 | 5 | 617 | |||
XM_017011695.1 | XP_016867184.1 | 2 | 228 | |||
XM_017011696.1 | XP_016867185.1 | 3 | 454 | |||
GenBank mRNA | AK090648.1 | - | 5 | 1260 | ||
BC007078.1 | - | 2 | 178 | |||
BC019234.1 | - | 2 | 182 | |||
BC063818.1 | - | 2 | 208 | 6503687 | ||
BC108740.1 | - | 2 | 173 | |||
DA807335.1 | - | 2 | 242 |