Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVIL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003930.2 | 3 | 1174 | |||
NR_110923.1 | 3 | 1200 | ||||
NR_110924.1 | 3 | 1199 | ||||
GenBank mRNA | BC031656.1 | - | 3 | 1129 | ||
BC107795.1 | - | 3 | 1183 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA570840.1 | - | 314 | Chr. - - [-] on Build |