Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027457.2 | 2 | 366 | |||
GenBank mRNA | BX161407.1 | - | 1 | 169 | ||
BX248748.1 | - | 2 | 338 | |||
BX356743.2 | - | 2 | 292 | |||
DB461309.1 | - | 2 | 356 |