Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024381.1 | 1 | 545 | |||
GenBank mRNA | AK057431.1 | - | 1 | 545 | ||
BC042984.2 | - | 1 | 513 | |||
BC104250.1 | - | 1 | 487 | |||
BC104251.1 | - | 1 | 484 | |||
BC113002.1 | - | 1 | 487 | |||
BC113003.1 | - | 1 | 487 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG721941.1 | - | 1 | 549 | Chr. 22 - 19564961 - 19566843 [-] on Build GRCh38 |