Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP8B5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003581.2 | 10 | 766 | |||
NR_003582.1 | 10 | 770 | ||||
NR_110320.1 | 10 | 766 | ||||
GenBank mRNA | AK310278.1 | - | 10 | 742 | ||
BC031276.1 | - | 10 | 753 | |||
BC035229.1 | - | 9 | 2329 | |||
BC130448.1 | - | 5 | 530 | |||
BC130450.1 | - | 5 | 530 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB461154.1 | - | 7 | 451 | Chr. 9 - 35406789 - 35447316 [+] on Build GRCh38 |