Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP8B5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003581.2 | 11 | 885 | |||
NR_003582.1 | 11 | 889 | ||||
NR_110320.1 | 11 | 885 | ||||
GenBank mRNA | AK310278.1 | - | 11 | 861 | ||
BC031276.1 | - | 11 | 872 | |||
BC130448.1 | - | 6 | 649 |