Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026829.1 | 1 | 1203 | |||
GenBank mRNA | AF085991.1 | - | 1 | 473 | ||
AF280797.2 | - | 1 | 1203 | |||
BC015602.2 | - | 1 | 1155 |