Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026838.1 | 4 | 758 | |||
NR_026839.1 | 3 | 661 | ||||
NR_026840.1 | 4 | 519 | ||||
NR_026841.1 | 4 | 501 | ||||
NR_026842.1 | 3 | 404 | ||||
GenBank mRNA | AF321193.1 | - | 4 | 476 | ||
AF426256.1 | - | 4 | 424 | |||
AJ001904.1 | - | 4 | 461 | |||
AJ297914.2 | - | 4 | 479 | |||
AJ301615.2 | - | 3 | 382 | |||
AJ305085.2 | - | 3 | 639 | |||
AJ306839.2 | - | 4 | 497 | |||
AJ306840.2 | - | 4 | 551 | |||
AJ783421.1 | - | 4 | 736 | |||
BC015981.1 | - | 3 | 386 | |||
BC029353.1 | - | 3 | 661 | |||
BC070229.1 | - | 3 | 363 | |||
BG189423.1 | - | 1 | 156 |