Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY3-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015359.1 | 4 | 721 | |||
GenBank mRNA | AY250185.1 | - | 7 | 1244 | ||
BC019264.1 | - | 7 | 797 | |||
BC033127.1 | - | 4 | 702 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL523811.3 | - | 4 | 721 | Chr. 4 - 84966818 - 85006497 [+] on Build GRCh38 |